Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020452.3 | NP_065185.1 | 19 - 20 | 2136 | 58 |
XM_005245355.2 | XP_005245412.1 | 18 - 19 | 2283 | 58 | |
XM_005245356.3 | XP_005245413.1 | 19 - 20 | 2236 | 58 | |
XM_017001871.1 | XP_016857360.1 | 18 - 19 | 2286 | 58 | |
XM_017001872.1 | XP_016857361.1 | 19 - 20 | 2235 | 58 | |
GenBank mRNA | AB032963.1 | - | 9 - 10 | 1268 | 58 |
AK304806.1 | - | 1 - 2 | 84 | 58 | |
AY302537.1 | - | 19 - 20 | 2136 | 58 | |
BC007837.2 | - | 5 - 6 | 754 | 58 | |
BC041573.1 | - | 5 - 6 | 754 | 58 |