Assay Design
Gene Transcripts
- Gene Symbol
- ATP8B2
- Entrez Gene ID
-
57198
- Gene Name
- ATPase phospholipid transporting 8B2
- Gene Aliases
-
ATPID
- Chromosome Location
-
Chr.1: 154325541 - 154351307 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK054886.1 |
- |
11 - 12 |
1273 |
83 |
View Full Product Details