Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318751.1 | NP_001305680.1 | 24 - 25 | 2435 | 75 |
NM_001318752.1 | NP_001305681.1 | 22 - 23 | 2316 | 75 | |
NM_001318753.1 | NP_001305682.1 | 7 - 8 | 482 | 75 | |
NM_001318754.1 | NP_001305683.1 | 7 - 8 | 482 | 75 | |
NM_030820.3 | NP_110447.2 | 23 - 24 | 2361 | 75 | |
NR_134849.1 | 23 - 24 | 2353 | 75 | ||
NR_134850.1 | 24 - 25 | 2378 | 75 | ||
XM_011514924.2 | XP_011513226.1 | 23 - 24 | 2304 | 75 | |
XM_011514925.2 | XP_011513227.1 | 25 - 26 | 2474 | 75 | |
XM_011514926.1 | XP_011513228.1 | 23 - 24 | 2232 | 75 | |
XM_011514927.1 | XP_011513229.1 | 23 - 24 | 2186 | 75 | |
GenBank mRNA | AF330693.1 | - | 22 - 23 | 2282 | 75 |
AF370383.1 | - | 11 - 12 | 617 | 75 | |
AF414088.1 | - | 23 - 24 | 2331 | 75 | |
AK096444.1 | - | 7 - 8 | 485 | 75 | |
AK126586.1 | - | 24 - 25 | 2246 | 75 | |
AK313398.1 | - | 23 - 24 | 2337 | 75 | |
AL136624.1 | - | 23 - 24 | 2331 | 75 | |
BC045597.1 | - | 23 - 24 | 2308 | 75 | |
BC126108.1 | - | 23 - 24 | 2216 | 75 | |
BC143862.1 | - | 23 - 24 | 2208 | 75 | |
BC143863.1 | - | 24 - 25 | 2233 | 75 | |
BC143865.1 | - | 24 - 25 | 2290 | 75 | |
DA826579.1 | - | 7 - 8 | 472 | 75 |