Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318751.1 | NP_001305680.1 | 4 - 5 | 940 | 120 |
NM_001318752.1 | NP_001305681.1 | 3 - 4 | 830 | 120 | |
NM_030820.3 | NP_110447.2 | 3 - 4 | 866 | 120 | |
NR_134849.1 | 3 - 4 | 866 | 120 | ||
NR_134850.1 | 3 - 4 | 866 | 120 | ||
NR_134851.1 | 3 - 4 | 866 | 120 | ||
XM_006715223.1 | XP_006715286.1 | 3 - 4 | 1032 | 120 | |
XM_011514924.2 | XP_011513226.1 | 3 - 4 | 809 | 120 | |
XM_011514925.2 | XP_011513227.1 | 5 - 6 | 979 | 120 | |
XM_011514926.1 | XP_011513228.1 | 3 - 4 | 737 | 120 | |
XM_011514927.1 | XP_011513229.1 | 3 - 4 | 691 | 120 | |
GenBank mRNA | AF330693.1 | - | 3 - 4 | 796 | 120 |
AF414088.1 | - | 3 - 4 | 836 | 120 | |
AK126586.1 | - | 3 - 4 | 732 | 120 | |
AK313398.1 | - | 3 - 4 | 842 | 120 | |
AL136624.1 | - | 3 - 4 | 836 | 120 | |
BC045597.1 | - | 3 - 4 | 821 | 120 | |
BC126108.1 | - | 3 - 4 | 721 | 120 | |
BC143862.1 | - | 3 - 4 | 721 | 120 | |
BC143863.1 | - | 3 - 4 | 721 | 120 | |
BC143864.1 | - | 3 - 4 | 721 | 120 | |
BC143865.1 | - | 4 - 5 | 795 | 120 |