Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289174.1 | NP_001276103.1 | 6 - 7 | 1385 | 68 |
NM_001289175.1 | NP_001276104.1 | 7 - 8 | 670 | 68 | |
NM_001289176.1 | NP_001276105.1 | 7 - 8 | 670 | 68 | |
NM_022553.5 | NP_072047.4 | 7 - 8 | 989 | 68 | |
XM_011514797.1 | XP_011513099.1 | 6 - 7 | 868 | 68 | |
XM_011514798.2 | XP_011513100.1 | 6 - 7 | 758 | 68 | |
XM_011514799.1 | XP_011513101.1 | 6 - 7 | 574 | 68 | |
XM_017011177.1 | XP_016866666.1 | 7 - 8 | 924 | 68 | |
XM_017011178.1 | XP_016866667.1 | 7 - 8 | 671 | 68 | |
XM_017011179.1 | XP_016866668.1 | 7 - 8 | 924 | 68 | |
GenBank mRNA | AK001725.1 | - | 7 - 8 | 667 | 68 |
AK093057.1 | - | 6 - 7 | 1296 | 68 | |
AK222867.1 | - | 7 - 8 | 715 | 68 | |
AK297935.1 | - | 7 - 8 | 645 | 68 | |
AK299563.1 | - | 8 - 9 | 825 | 68 | |
AK300308.1 | - | 7 - 8 | 930 | 68 | |
AK300430.1 | - | 7 - 8 | 687 | 68 | |
AK304080.1 | - | 7 - 8 | 893 | 68 | |
AL559461.3 | - | 7 - 8 | 675 | 68 | |
AX775737.1 | - | 7 - 8 | 833 | 68 | |
BC032108.1 | - | 7 - 8 | 851 | 68 | |
BC040114.1 | - | 7 - 8 | 870 | 68 |