Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289174.1 | NP_001276103.1 | 8 - 9 | 1618 | 96 |
NM_001289175.1 | NP_001276104.1 | 9 - 10 | 903 | 96 | |
NM_022553.5 | NP_072047.4 | 9 - 10 | 1222 | 96 | |
XM_011514797.1 | XP_011513099.1 | 8 - 9 | 1101 | 96 | |
XM_011514798.2 | XP_011513100.1 | 8 - 9 | 991 | 96 | |
XM_011514799.1 | XP_011513101.1 | 8 - 9 | 807 | 96 | |
XM_017011177.1 | XP_016866666.1 | 9 - 10 | 1157 | 96 | |
XM_017011178.1 | XP_016866667.1 | 9 - 10 | 904 | 96 | |
XM_017011179.1 | XP_016866668.1 | 9 - 10 | 1157 | 96 | |
GenBank mRNA | AK093057.1 | - | 8 - 9 | 1529 | 96 |
AK222867.1 | - | 9 - 10 | 948 | 96 | |
AK297935.1 | - | 9 - 10 | 878 | 96 | |
AK299563.1 | - | 10 - 11 | 1058 | 96 | |
AK300430.1 | - | 9 - 10 | 920 | 96 | |
AK304080.1 | - | 9 - 10 | 1126 | 96 | |
AL390171.1 | - | 2 - 3 | 171 | 96 | |
AX775737.1 | - | 9 - 10 | 1066 | 96 | |
BC032108.1 | - | 9 - 10 | 1084 | 96 | |
BC040114.1 | - | 9 - 10 | 1103 | 96 |