Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128610.2 | NP_001122082.1 | 2 - 3 | 453 | 154 |
NM_005154.4 | NP_005145.3 | 2 - 3 | 313 | 154 | |
XM_006720761.3 | XP_006720824.1 | 3 - 4 | 633 | 154 | |
XM_006720762.3 | XP_006720825.1 | 3 - 4 | 633 | 154 | |
XM_011522193.2 | XP_011520495.1 | 3 - 4 | 470 | 154 | |
XM_017022718.1 | XP_016878207.1 | 2 - 3 | 308 | 154 | |
XM_017022719.1 | XP_016878208.1 | 2 - 3 | 425 | 154 | |
XM_017022720.1 | XP_016878209.1 | 3 - 4 | 470 | 154 | |
XM_017022721.1 | XP_016878210.1 | 2 - 3 | 296 | 154 | |
GenBank mRNA | AK292400.1 | - | 2 - 3 | 308 | 154 |
BC015545.1 | - | 3 - 4 | 587 | 154 | |
BC026954.1 | - | 2 - 3 | 403 | 154 | |
BC038801.1 | - | 2 - 3 | 301 | 154 | |
BC051345.1 | - | 2 - 3 | 352 | 154 | |
BC062694.1 | - | 1 - 2 | 144 | 154 | |
BC092443.1 | - | 2 - 3 | 275 | 154 | |
BC110590.2 | - | 2 - 3 | 279 | 154 | |
BX537420.1 | - | 2 - 3 | 308 | 154 | |
D29956.1 | - | 2 - 3 | 427 | 154 | |
DA112964.1 | - | 2 - 3 | 314 | 154 |