Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005423.4 | NP_005414.1 | 1 - 2 | 227 | 101 |
GenBank mRNA | AA307808.1 | - | 1 - 2 | 227 | 101 |
BC032820.1 | - | 1 - 2 | 106 | 101 | |
BM817851.1 | - | 1 - 2 | 126 | 101 | |
BM853946.1 | - | 1 - 2 | 124 | 101 | |
X51698.1 | - | 1 - 2 | 90 | 100 |