Assay Design
Gene Transcripts
- Gene Symbol
- GJD3
- Entrez Gene ID
-
125111
- Gene Name
- gap junction protein delta 3
- Gene Aliases
-
CX31.9, Cx30.2, GJA11, GJC1
- Chromosome Location
-
Chr.17: 40360652 - 40364693 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_152219.3 |
NP_689343.3 |
1 - 1 |
2218 |
78 |
View Full Product Details