Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004719.2 | NP_004710.2 | 3 - 4 | 504 | 150 |
XM_005269230.2 | XP_005269287.2 | 3 - 4 | 394 | 150 | |
XM_011538984.2 | XP_011537286.1 | 4 - 5 | 2023 | 150 | |
XM_011538985.1 | XP_011537287.1 | 3 - 4 | 338 | 150 | |
GenBank mRNA | AF030234.1 | - | 3 - 4 | 452 | 151 |
AK096682.1 | - | 4 - 5 | 1995 | 150 | |
AK302999.1 | - | 3 - 4 | 429 | 150 |