Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 11 - 12 | 1159 | 109 |
NM_018482.3 | NP_060952.2 | 10 - 11 | 1057 | 109 | |
XM_005250925.1 | XP_005250982.1 | 10 - 11 | 1057 | 109 | |
XM_006716563.3 | XP_006716626.1 | 10 - 11 | 876 | 109 | |
XM_006716564.1 | XP_006716627.1 | 11 - 12 | 1035 | 109 | |
XM_006716565.3 | XP_006716628.1 | 9 - 10 | 872 | 109 | |
XM_006716566.1 | XP_006716629.1 | 10 - 11 | 1057 | 109 | |
XM_011517052.2 | XP_011515354.1 | 10 - 11 | 873 | 109 | |
XM_011517053.1 | XP_011515355.1 | 10 - 11 | 1079 | 109 | |
XM_017013467.1 | XP_016868956.1 | 8 - 9 | 894 | 109 | |
XM_017013468.1 | XP_016868957.1 | 9 - 10 | 1118 | 109 | |
XM_017013469.1 | XP_016868958.1 | 10 - 11 | 1057 | 109 | |
GenBank mRNA | AB033075.1 | - | 3 - 4 | 281 | 109 |
BC137135.1 | - | 9 - 10 | 854 | 109 | |
BC144493.1 | - | 10 - 11 | 956 | 109 | |
BX537768.1 | - | 3 - 4 | 310 | 109 |