Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 26 - 27 | 2901 | 121 |
NM_018482.3 | NP_060952.2 | 25 - 26 | 2799 | 121 | |
XM_005250925.1 | XP_005250982.1 | 26 - 27 | 2808 | 121 | |
XM_006716563.3 | XP_006716626.1 | 26 - 27 | 2627 | 121 | |
XM_006716564.1 | XP_006716627.1 | 27 - 28 | 2786 | 121 | |
XM_006716565.3 | XP_006716628.1 | 25 - 26 | 2623 | 121 | |
XM_011517052.2 | XP_011515354.1 | 26 - 27 | 2624 | 121 | |
XM_011517053.1 | XP_011515355.1 | 26 - 27 | 2830 | 121 | |
XM_017013467.1 | XP_016868956.1 | 24 - 25 | 2645 | 121 | |
XM_017013468.1 | XP_016868957.1 | 25 - 26 | 2869 | 121 | |
GenBank mRNA | AB033075.1 | - | 19 - 20 | 2032 | 121 |
BC137135.1 | - | 24 - 25 | 2596 | 121 | |
BC144493.1 | - | 25 - 26 | 2698 | 121 | |
BX537768.1 | - | 18 - 19 | 2052 | 121 |