Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 4 - 5 | 516 | 82 |
NM_018482.3 | NP_060952.2 | 3 - 4 | 414 | 82 | |
XM_005250925.1 | XP_005250982.1 | 3 - 4 | 414 | 82 | |
XM_006716563.3 | XP_006716626.1 | 3 - 4 | 233 | 82 | |
XM_006716564.1 | XP_006716627.1 | 4 - 5 | 392 | 82 | |
XM_006716566.1 | XP_006716629.1 | 3 - 4 | 414 | 82 | |
XM_011517052.2 | XP_011515354.1 | 3 - 4 | 230 | 82 | |
XM_011517053.1 | XP_011515355.1 | 3 - 4 | 436 | 82 | |
XM_017013469.1 | XP_016868958.1 | 3 - 4 | 414 | 82 | |
GenBank mRNA | AW236958.1 | - | 4 - 5 | 82 | |
BC137135.1 | - | 2 - 3 | 211 | 82 | |
BC144493.1 | - | 3 - 4 | 313 | 82 | |
DA319233.1 | - | 3 - 4 | 414 | 82 |