Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 8 - 9 | 865 | 78 |
NM_018482.3 | NP_060952.2 | 7 - 8 | 763 | 78 | |
XM_005250925.1 | XP_005250982.1 | 7 - 8 | 763 | 78 | |
XM_006716563.3 | XP_006716626.1 | 7 - 8 | 582 | 78 | |
XM_006716564.1 | XP_006716627.1 | 8 - 9 | 741 | 78 | |
XM_006716565.3 | XP_006716628.1 | 6 - 7 | 578 | 78 | |
XM_006716566.1 | XP_006716629.1 | 7 - 8 | 763 | 78 | |
XM_011517052.2 | XP_011515354.1 | 7 - 8 | 579 | 78 | |
XM_011517053.1 | XP_011515355.1 | 7 - 8 | 785 | 78 | |
XM_017013467.1 | XP_016868956.1 | 5 - 6 | 600 | 78 | |
XM_017013468.1 | XP_016868957.1 | 6 - 7 | 824 | 78 | |
XM_017013469.1 | XP_016868958.1 | 7 - 8 | 763 | 78 | |
GenBank mRNA | BC137135.1 | - | 6 - 7 | 560 | 78 |
BC144493.1 | - | 7 - 8 | 662 | 78 |