Assay Design



Gene Transcripts

Gene Symbol
LANCL1
Entrez Gene ID
10314
Gene Name
LanC like 1
Gene Aliases
GPR69A, p40
Chromosome Location
Chr.2: 210431249 - 210477618 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005246243.2 XP_005246300.1 1 - 2 172 66

View Full Product Details