Assay Design



Gene Transcripts

Gene Symbol
RASAL1
Entrez Gene ID
8437
Gene Name
RAS protein activator like 1
Gene Aliases
RASAL
Chromosome Location
Chr.12: 113096515 - 113136239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193521.1 NP_001180450.1 16 - 17 2119 127
GenBank mRNA BC014420.1 - 16 - 17 2010 127

View Full Product Details