Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193520.1 | NP_001180449.1 | 9 - 10 | 1035 | 81 |
NM_001193521.1 | NP_001180450.1 | 8 - 9 | 1026 | 81 | |
NM_001301202.1 | NP_001288131.1 | 8 - 9 | 871 | 81 | |
NM_004658.2 | NP_004649.2 | 9 - 10 | 1050 | 81 | |
XM_005253950.4 | XP_005254007.1 | 9 - 10 | 1050 | 81 | |
XM_006719641.3 | XP_006719704.1 | 9 - 10 | 889 | 81 | |
XM_006719642.3 | XP_006719705.1 | 8 - 9 | 1022 | 81 | |
XM_011538852.2 | XP_011537154.1 | 8 - 9 | 1022 | 81 | |
XM_011538853.2 | XP_011537155.1 | 8 - 9 | 1022 | 81 | |
XM_017020028.1 | XP_016875517.1 | 9 - 10 | 1050 | 81 | |
XM_017020029.1 | XP_016875518.1 | 8 - 9 | 1022 | 81 | |
XM_017020030.1 | XP_016875519.1 | 9 - 10 | 1050 | 81 | |
XM_017020031.1 | XP_016875520.1 | 9 - 10 | 1050 | 81 | |
GenBank mRNA | AB208935.1 | - | 9 - 10 | 1020 | 81 |
AF086713.1 | - | 9 - 10 | 1027 | 81 | |
AK294350.1 | - | 9 - 10 | 1050 | 81 | |
AL136672.1 | - | 8 - 9 | 967 | 81 | |
BC014420.1 | - | 8 - 9 | 917 | 81 | |
BC093724.1 | - | 8 - 9 | 776 | 81 | |
BC143261.1 | - | 8 - 9 | 776 | 81 |