Assay Design



Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 12 - 13 1159 67
NM_001190442.1 NP_001177371.1 13 - 14 1031 67
NM_001284212.2 NP_001271141.1 10 - 11 810 67
NM_001284213.2 NP_001271142.1 9 - 10 717 67
NM_173060.3 NP_775083.1 11 - 12 867 67
XM_006714696.3 XP_006714759.1 15 - 16 1173 67
XM_006714697.3 XP_006714760.1 14 - 15 1190 67
XM_006714698.3 XP_006714761.1 13 - 14 1151 67
XM_006714699.3 XP_006714762.1 14 - 15 1114 67
XM_006714700.3 XP_006714763.1 13 - 14 1133 67
XM_006714701.3 XP_006714764.1 13 - 14 1149 67
XM_006714702.3 XP_006714765.1 13 - 14 1025 67
XM_006714703.3 XP_006714766.1 12 - 13 1110 67
XM_006714704.3 XP_006714767.1 13 - 14 1058 67
XM_006714705.3 XP_006714768.1 12 - 13 1067 67
XM_006714706.3 XP_006714769.1 12 - 13 985 67
XM_006714707.3 XP_006714770.1 12 - 13 936 67
XM_006714708.3 XP_006714771.1 11 - 12 896 67
XM_006714709.3 XP_006714772.1 11 - 12 859 67
XM_006714710.3 XP_006714773.1 10 - 11 814 67
XM_006714711.3 XP_006714774.1 10 - 11 859 67
XM_006714712.3 XP_006714775.1 9 - 10 806 67
XM_011543654.2 XP_011541956.1 13 - 14 1109 67
XM_011543655.2 XP_011541957.1 14 - 15 1190 67
XM_011543656.2 XP_011541958.1 12 - 13 1053 67
XM_011543657.2 XP_011541959.1 12 - 13 935 67
XM_011543658.2 XP_011541960.1 10 - 11 788 67
XM_017009911.1 XP_016865400.1 13 - 14 1151 67
XM_017009912.1 XP_016865401.1 14 - 15 1162 67
XM_017009913.1 XP_016865402.1 14 - 15 1131 67
XM_017009914.1 XP_016865403.1 13 - 14 1133 67
XM_017009915.1 XP_016865404.1 14 - 15 1135 67
XM_017009916.1 XP_016865405.1 13 - 14 1149 67
XM_017009917.1 XP_016865406.1 13 - 14 1067 67
XM_017009918.1 XP_016865407.1 12 - 13 1094 67
XM_017009919.1 XP_016865408.1 13 - 14 1151 67
XM_017009920.1 XP_016865409.1 12 - 13 977 67
XM_017009921.1 XP_016865410.1 12 - 13 994 67
XM_017009922.1 XP_016865411.1 11 - 12 1028 67
XM_017009923.1 XP_016865412.1 11 - 12 1028 67
XM_017009924.1 XP_016865413.1 12 - 13 1094 67
XM_017009925.1 XP_016865414.1 11 - 12 946 67
XM_017009926.1 XP_016865415.1 11 - 12 946 67
XM_017009927.1 XP_016865416.1 12 - 13 935 67
XM_017009928.1 XP_016865417.1 11 - 12 934 67
XM_017009929.1 XP_016865418.1 11 - 12 853 67
XM_017009930.1 XP_016865419.1 10 - 11 820 67
XM_017009931.1 XP_016865420.1 10 - 11 814 67
XM_017009932.1 XP_016865421.1 9 - 10 877 67
GenBank mRNA AB208816.1 - 13 - 14 995 67
AF327443.1 - 8 - 9 709 67
AK296919.1 - 13 - 14 989 67
AK297879.1 - 11 - 12 809 67
AK298541.1 - 12 - 13 936 67
AK299384.1 - 12 - 13 840 67
AK300624.1 - 9 - 10 700 67
AK303836.1 - 5 - 6 390 67
AK314290.1 - 10 - 11 812 67
AK315146.1 - 10 - 11 811 67
AL832174.1 - 11 - 12 852 67
AL832349.1 - 11 - 12 855 67
BC013579.1 - 12 - 13 862 67
BC016066.1 - 12 - 13 1131 67
BT009783.1 - 9 - 10 644 67
BX648397.1 - 10 - 11 761 67
D16217.1 - 12 - 13 929 67
M33328.1 - 4 - 5 356 67
U26724.2 - 3 - 4 292 67
U31345.1 - 1 - 2 39 67

View Full Product Details