Assay Design



Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 20 - 21 1851 93
NM_001190442.1 NP_001177371.1 21 - 22 1723 93
NM_001284212.2 NP_001271141.1 18 - 19 1502 93
NM_001284213.2 NP_001271142.1 17 - 18 1409 93
NM_173060.3 NP_775083.1 19 - 20 1559 93
NR_104285.1 7 - 8 715 93
XM_006714696.3 XP_006714759.1 23 - 24 1865 93
XM_006714697.3 XP_006714760.1 22 - 23 1882 93
XM_006714698.3 XP_006714761.1 21 - 22 1843 93
XM_006714699.3 XP_006714762.1 22 - 23 1806 93
XM_006714700.3 XP_006714763.1 21 - 22 1825 93
XM_006714701.3 XP_006714764.1 21 - 22 1841 93
XM_006714702.3 XP_006714765.1 21 - 22 1717 93
XM_006714703.3 XP_006714766.1 20 - 21 1802 93
XM_006714704.3 XP_006714767.1 21 - 22 1750 93
XM_006714705.3 XP_006714768.1 20 - 21 1759 93
XM_006714706.3 XP_006714769.1 20 - 21 1677 93
XM_006714707.3 XP_006714770.1 20 - 21 1628 93
XM_006714708.3 XP_006714771.1 19 - 20 1588 93
XM_006714709.3 XP_006714772.1 19 - 20 1551 93
XM_006714710.3 XP_006714773.1 18 - 19 1506 93
XM_006714711.3 XP_006714774.1 18 - 19 1551 93
XM_006714712.3 XP_006714775.1 17 - 18 1498 93
XM_006714713.3 XP_006714776.1 7 - 8 715 93
XM_006714714.3 XP_006714777.1 7 - 8 715 93
XM_006714715.3 XP_006714778.1 8 - 9 1441 93
XM_011543654.2 XP_011541956.1 21 - 22 1801 93
XM_011543655.2 XP_011541957.1 21 - 22 1798 93
XM_011543656.2 XP_011541958.1 20 - 21 1745 93
XM_011543657.2 XP_011541959.1 20 - 21 1627 93
XM_011543658.2 XP_011541960.1 18 - 19 1480 93
XM_017009911.1 XP_016865400.1 21 - 22 1843 93
XM_017009912.1 XP_016865401.1 22 - 23 1854 93
XM_017009913.1 XP_016865402.1 22 - 23 1823 93
XM_017009914.1 XP_016865403.1 21 - 22 1825 93
XM_017009915.1 XP_016865404.1 22 - 23 1827 93
XM_017009916.1 XP_016865405.1 21 - 22 1841 93
XM_017009917.1 XP_016865406.1 21 - 22 1759 93
XM_017009918.1 XP_016865407.1 20 - 21 1786 93
XM_017009919.1 XP_016865408.1 20 - 21 1759 93
XM_017009920.1 XP_016865409.1 20 - 21 1669 93
XM_017009921.1 XP_016865410.1 20 - 21 1686 93
XM_017009922.1 XP_016865411.1 19 - 20 1720 93
XM_017009923.1 XP_016865412.1 19 - 20 1720 93
XM_017009924.1 XP_016865413.1 19 - 20 1702 93
XM_017009925.1 XP_016865414.1 19 - 20 1638 93
XM_017009926.1 XP_016865415.1 19 - 20 1638 93
XM_017009927.1 XP_016865416.1 20 - 21 1627 93
XM_017009928.1 XP_016865417.1 19 - 20 1626 93
XM_017009929.1 XP_016865418.1 19 - 20 1545 93
XM_017009930.1 XP_016865419.1 18 - 19 1512 93
XM_017009931.1 XP_016865420.1 18 - 19 1506 93
XM_017009932.1 XP_016865421.1 17 - 18 1569 93
GenBank mRNA AB208816.1 - 21 - 22 1687 93
AF327443.1 - 16 - 17 1401 93
AK292289.1 - 7 - 8 715 93
AK292356.1 - 7 - 8 688 93
AK296919.1 - 21 - 22 1681 93
AK297879.1 - 19 - 20 1501 93
AK298541.1 - 20 - 21 1628 93
AK299402.1 - 20 - 21 1608 93
AK300624.1 - 17 - 18 1392 93
AK302206.1 - 8 - 9 715 93
AK311585.1 - 8 - 9 893 93
AK311598.1 - 8 - 9 769 93
AK314290.1 - 18 - 19 1504 93
AK315146.1 - 18 - 19 1503 93
AK315673.1 - 7 - 8 663 93
AL832174.1 - 19 - 20 1544 93
AL832349.1 - 19 - 20 1547 93
AY189741.1 - 9 - 10 714 93
BC013579.1 - 20 - 21 1554 93
BC078149.1 - 15 - 16 1137 93
BT009783.1 - 17 - 18 1336 93
BX648397.1 - 18 - 19 1453 93
D16217.1 - 20 - 21 1621 93
U31345.1 - 9 - 10 731 93
U38525.1 - 2 - 3 187 93

View Full Product Details