Assay Design



Gene Transcripts

Gene Symbol
SYT1
Entrez Gene ID
6857
Gene Name
synaptotagmin 1
Gene Aliases
P65, SVP65, SYT
Chromosome Location
Chr.12: 78863993 - 79452008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291901.1 NP_001278830.1 2 - 3 574 66
XM_011538710.1 XP_011537012.1 2 - 3 573 66
XM_017019909.1 XP_016875398.1 2 - 3 233 66

View Full Product Details