Assay Design



Gene Transcripts

Gene Symbol
SYT1
Entrez Gene ID
6857
Gene Name
synaptotagmin 1
Gene Aliases
P65, SVP65, SYT
Chromosome Location
Chr.12: 78863993 - 79452008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291901.1 NP_001278830.1 4 - 5 942 132
XM_005269113.2 XP_005269170.1 5 - 6 1007 132
XM_006719576.1 XP_006719639.1 4 - 5 630 132

View Full Product Details