Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135805.1 | NP_001129277.1 | 8 - 9 | 1314 | 81 |
NM_001135806.1 | NP_001129278.1 | 6 - 7 | 900 | 81 | |
NM_001291901.1 | NP_001278830.1 | 6 - 7 | 1225 | 81 | |
NM_005639.2 | NP_005630.1 | 7 - 8 | 1300 | 81 | |
XM_005269113.2 | XP_005269170.1 | 7 - 8 | 1290 | 81 | |
XM_006719576.1 | XP_006719639.1 | 6 - 7 | 913 | 81 | |
XM_011538710.1 | XP_011537012.1 | 6 - 7 | 1233 | 81 | |
XM_017019909.1 | XP_016875398.1 | 6 - 7 | 893 | 81 | |
GenBank mRNA | AK094616.1 | - | 8 - 9 | 1314 | 81 |
AK126908.1 | - | 7 - 8 | 1300 | 81 | |
BC058917.1 | - | 7 - 8 | 908 | 81 | |
BX647240.1 | - | 7 - 8 | 1194 | 81 | |
CR627387.1 | - | 6 - 7 | 900 | 81 | |
M55047.1 | - | 4 - 5 | 670 | 81 |