Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135805.1 | NP_001129277.1 | 10 - 11 | 1599 | 84 |
NM_001135806.1 | NP_001129278.1 | 8 - 9 | 1185 | 84 | |
NM_001291901.1 | NP_001278830.1 | 8 - 9 | 1510 | 84 | |
NM_005639.2 | NP_005630.1 | 9 - 10 | 1585 | 84 | |
XM_005269113.2 | XP_005269170.1 | 9 - 10 | 1575 | 84 | |
XM_006719576.1 | XP_006719639.1 | 8 - 9 | 1198 | 84 | |
XM_011538710.1 | XP_011537012.1 | 8 - 9 | 1518 | 84 | |
XM_017019909.1 | XP_016875398.1 | 8 - 9 | 1178 | 84 | |
GenBank mRNA | AK094616.1 | - | 10 - 11 | 1599 | 84 |
AK126908.1 | - | 9 - 10 | 1585 | 84 | |
AK308743.1 | - | 6 - 7 | 701 | 84 | |
BC058917.1 | - | 9 - 10 | 1193 | 84 | |
BX647240.1 | - | 9 - 10 | 1479 | 84 | |
CR627387.1 | - | 8 - 9 | 1185 | 84 | |
M55047.1 | - | 6 - 7 | 955 | 84 |