Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301778.1 | NP_001288707.1 | 8 - 9 | 1029 | 54 |
NM_198935.2 | NP_945173.1 | 7 - 8 | 924 | 54 | |
NR_125980.1 | 8 - 9 | 1303 | 54 | ||
NR_125981.1 | 7 - 8 | 924 | 54 | ||
XM_005260389.3 | XP_005260446.1 | 7 - 8 | 879 | 54 | |
XM_005260391.1 | XP_005260448.1 | 8 - 9 | 1063 | 54 | |
XM_011528764.2 | XP_011527066.1 | 8 - 9 | 2055 | 54 | |
XM_011528767.1 | XP_011527069.1 | 6 - 7 | 865 | 54 | |
XM_017027784.1 | XP_016883273.1 | 7 - 8 | 1245 | 54 | |
GenBank mRNA | AB014593.1 | - | 7 - 8 | 849 | 54 |
AK125656.1 | - | 8 - 9 | 996 | 54 | |
AK291485.1 | - | 7 - 8 | 899 | 54 | |
AK299881.1 | - | 7 - 8 | 899 | 54 | |
AY203931.1 | - | 4 - 5 | 611 | 54 | |
BC034494.2 | - | 7 - 8 | 878 | 54 | |
BC068993.1 | - | 8 - 9 | 1241 | 54 | |
BX640848.1 | - | 8 - 9 | 1061 | 54 |