Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195518.1 | NP_001182447.1 | 11 - 12 | 1457 | 81 |
NM_001195519.1 | NP_001182448.1 | 6 - 7 | 807 | 81 | |
NM_006077.3 | NP_006068.2 | 12 - 13 | 1463 | 81 | |
XM_005269383.2 | XP_005269440.1 | 11 - 12 | 1379 | 81 | |
XM_005269386.2 | XP_005269443.1 | 7 - 8 | 920 | 81 | |
XM_011539119.1 | XP_011537421.1 | 12 - 13 | 1529 | 81 | |
GenBank mRNA | AK022697.1 | - | 9 - 10 | 922 | 81 |
AK023318.1 | - | 11 - 12 | 1376 | 81 | |
AK225298.1 | - | 11 - 12 | 1361 | 81 | |
AK225695.1 | - | 11 - 12 | 1345 | 81 | |
AK296086.1 | - | 6 - 7 | 774 | 81 | |
AK298347.1 | - | 8 - 9 | 983 | 81 | |
AL117423.1 | - | 12 - 13 | 1354 | 81 | |
BC004190.2 | - | 11 - 12 | 1369 | 81 | |
BC004216.2 | - | 11 - 12 | 1369 | 81 | |
BC016641.1 | - | 11 - 12 | 1332 | 81 | |
Y17711.1 | - | 7 - 8 | 785 | 81 |