Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195518.1 | NP_001182447.1 | 9 - 10 | 1250 | 63 |
NM_001195519.1 | NP_001182448.1 | 4 - 5 | 600 | 63 | |
NM_006077.3 | NP_006068.2 | 10 - 11 | 1256 | 63 | |
XM_005269383.2 | XP_005269440.1 | 9 - 10 | 1172 | 63 | |
XM_005269386.2 | XP_005269443.1 | 5 - 6 | 713 | 63 | |
XM_011539119.1 | XP_011537421.1 | 10 - 11 | 1322 | 63 | |
GenBank mRNA | AK022697.1 | - | 6 - 7 | 595 | 63 |
AK023318.1 | - | 9 - 10 | 1169 | 63 | |
AK225298.1 | - | 9 - 10 | 1154 | 63 | |
AK225695.1 | - | 9 - 10 | 1138 | 63 | |
AK296086.1 | - | 4 - 5 | 567 | 63 | |
AK298347.1 | - | 6 - 7 | 776 | 63 | |
AL117423.1 | - | 10 - 11 | 1147 | 63 | |
BC004190.2 | - | 9 - 10 | 1162 | 63 | |
BC004216.2 | - | 9 - 10 | 1162 | 63 | |
BC016641.1 | - | 9 - 10 | 1125 | 63 | |
Y17711.1 | - | 5 - 6 | 578 | 63 |