Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195518.1 | NP_001182447.1 | 10 - 11 | 1369 | 73 |
NM_001195519.1 | NP_001182448.1 | 5 - 6 | 719 | 73 | |
NM_006077.3 | NP_006068.2 | 11 - 12 | 1375 | 73 | |
XM_005269383.2 | XP_005269440.1 | 10 - 11 | 1291 | 73 | |
XM_005269386.2 | XP_005269443.1 | 6 - 7 | 832 | 73 | |
XM_011539119.1 | XP_011537421.1 | 11 - 12 | 1441 | 73 | |
GenBank mRNA | AK023318.1 | - | 10 - 11 | 1288 | 73 |
AK225298.1 | - | 10 - 11 | 1273 | 73 | |
AK225695.1 | - | 10 - 11 | 1257 | 73 | |
AK296086.1 | - | 5 - 6 | 686 | 73 | |
AK298347.1 | - | 7 - 8 | 895 | 73 | |
AL117423.1 | - | 11 - 12 | 1266 | 73 | |
BC004190.2 | - | 10 - 11 | 1281 | 73 | |
BC004216.2 | - | 10 - 11 | 1281 | 73 | |
BC016641.1 | - | 10 - 11 | 1244 | 73 | |
Y17711.1 | - | 6 - 7 | 697 | 73 |