Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195518.1 | NP_001182447.1 | 7 - 8 | 917 | 125 |
NM_006077.3 | NP_006068.2 | 8 - 9 | 923 | 125 | |
XM_005269383.2 | XP_005269440.1 | 7 - 8 | 839 | 125 | |
XM_005269386.2 | XP_005269443.1 | 3 - 4 | 380 | 125 | |
XM_011539119.1 | XP_011537421.1 | 8 - 9 | 989 | 125 | |
GenBank mRNA | AK022697.1 | - | 4 - 5 | 262 | 125 |
AK023318.1 | - | 7 - 8 | 836 | 125 | |
AK225298.1 | - | 7 - 8 | 821 | 125 | |
AK225695.1 | - | 7 - 8 | 805 | 125 | |
AL117423.1 | - | 8 - 9 | 814 | 125 | |
BC004190.2 | - | 7 - 8 | 829 | 125 | |
BC004216.2 | - | 7 - 8 | 829 | 125 | |
BC016641.1 | - | 7 - 8 | 792 | 125 | |
Y17711.1 | - | 3 - 4 | 245 | 125 |