Assay Design



Gene Transcripts

Gene Symbol
DUOXA1
Entrez Gene ID
90527
Gene Name
dual oxidase maturation factor 1
Gene Aliases
NIP, NUMBIP, mol
Chromosome Location
Chr.15: 45117366 - 45129938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276267.1 NP_001263196.1 3 - 4 300 62
XM_011522183.2 XP_011520485.1 3 - 4 353 62
GenBank mRNA BC020841.1 - 3 - 4 300 62

View Full Product Details