Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002118.4 | NP_002109.2 | 3 - 4 | 882 | 77 |
GenBank mRNA | AB362554.1 | - | 1 - 2 | 86 | 77 |
AY645722.1 | - | 3 - 4 | 852 | 77 | |
BC027175.1 | - | 3 - 4 | 763 | 77 | |
CD742571.1 | - | 3 - 4 | 77 | ||
CR407606.1 | - | 3 - 4 | 619 | 77 | |
EL948395.1 | - | 2 - 3 | 315 | 77 | |
U15085.1 | - | 3 - 4 | 852 | 77 | |
Z23139.1 | - | 3 - 4 | 787 | 77 |