Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002118.4 | NP_002109.2 | 4 - 5 | 1000 | 89 |
GenBank mRNA | AY645722.1 | - | 4 - 5 | 970 | 89 |
BC027175.1 | - | 4 - 5 | 881 | 89 | |
CD742571.1 | - | 2 - 3 | 89 | ||
CR407606.1 | - | 4 - 5 | 737 | 89 | |
EL948395.1 | - | 3 - 4 | 433 | 89 | |
U15085.1 | - | 4 - 5 | 970 | 89 | |
Z23139.1 | - | 4 - 5 | 905 | 89 |