Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153610.4 | NP_705838.3 | 2 - 3 | 10705 | 126 |
XM_017009212.1 | XP_016864701.1 | 2 - 3 | 10573 | 126 | |
GenBank mRNA | AF533705.1 | - | 1 - 2 | 1299 | 126 |
AK123466.1 | - | 1 - 2 | 2063 | 126 | |
AL831986.1 | - | 1 - 2 | 1257 | 126 | |
AL832021.1 | - | 1 - 2 | 3079 | 126 | |
AL832368.1 | - | 1 - 2 | 2312 | 126 | |
AL834252.2 | - | 1 - 2 | 2422 | 126 | |
BC111529.2 | - | 1 - 2 | 2015 | 126 | |
BC111530.2 | - | 1 - 2 | 2015 | 126 | |
BX647628.1 | - | 1 - 2 | 2409 | 126 | |
BX647646.1 | - | 1 - 2 | 2409 | 126 |