Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153610.4 | NP_705838.3 | 3 - 4 | 10809 | 138 |
XM_017009212.1 | XP_016864701.1 | 3 - 4 | 10677 | 138 | |
GenBank mRNA | AF533705.1 | - | 2 - 3 | 1403 | 138 |
AL831966.1 | - | 2 - 3 | 138 | ||
AL831986.1 | - | 2 - 3 | 1361 | 138 | |
AL832021.1 | - | 2 - 3 | 3183 | 138 | |
AL832368.1 | - | 2 - 3 | 2416 | 138 | |
AL834252.2 | - | 2 - 3 | 2526 | 138 | |
BC111529.2 | - | 2 - 3 | 2119 | 138 | |
BC111530.2 | - | 2 - 3 | 138 | ||
BX647628.1 | - | 2 - 3 | 2513 | 138 | |
BX647646.1 | - | 2 - 3 | 2513 | 138 |