Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282541.1 | NP_001269470.1 | 19 - 20 | 1882 | 100 |
NM_001282542.1 | NP_001269471.1 | 19 - 20 | 1882 | 100 | |
NM_003176.3 | NP_003167.2 | 19 - 20 | 1879 | 100 | |
NR_104211.1 | 19 - 20 | 1879 | 100 | ||
XM_005271155.3 | XP_005271212.1 | 18 - 19 | 2203 | 100 | |
XM_005271156.1 | XP_005271213.1 | 18 - 19 | 1677 | 100 | |
XM_006710859.1 | XP_006710922.1 | 19 - 20 | 1790 | 100 | |
XM_011542037.2 | XP_011540339.1 | 19 - 20 | 1846 | 100 | |
XM_011542038.2 | XP_011540340.1 | 12 - 13 | 1073 | 100 | |
XM_017002184.1 | XP_016857673.1 | 19 - 20 | 1682 | 100 | |
XM_017002185.1 | XP_016857674.1 | 13 - 14 | 1108 | 100 | |
GenBank mRNA | AF273043.1 | - | 19 - 20 | 1817 | 100 |
AK315268.1 | - | 19 - 20 | 1841 | 100 | |
BC126266.1 | - | 19 - 20 | 1756 | 100 | |
BC144019.1 | - | 19 - 20 | 1759 | 100 | |
X95654.1 | - | 19 - 20 | 1733 | 100 |