Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310338.1 | NP_001297267.1 | 1 - 2 | 250 | 65 |
NM_001310339.1 | NP_001297268.1 | 1 - 2 | 250 | 65 | |
NM_052865.3 | NP_443097.1 | 1 - 2 | 250 | 65 | |
XM_005260870.3 | XP_005260927.1 | 1 - 2 | 198 | 65 | |
XM_006723663.3 | XP_006723726.1 | 1 - 2 | 198 | 65 | |
XM_011529395.1 | XP_011527697.1 | 1 - 2 | 250 | 65 | |
GenBank mRNA | AF306684.1 | - | 1 - 2 | 58 | 65 |
AF306685.1 | - | 1 - 2 | 58 | 65 | |
AF306686.1 | - | 1 - 2 | 96 | 65 | |
AK027503.1 | - | 1 - 2 | 54 | 65 | |
AK315532.1 | - | 1 - 2 | 86 | 65 | |
CV807140.1 | - | 1 - 2 | 63 | 65 | |
DB014120.1 | - | 1 - 2 | 250 | 65 |