Assay Design



Gene Transcripts

Gene Symbol
CSTF1
Entrez Gene ID
1477
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
CstF-50, CstFp50
Chromosome Location
Chr.20: 56392371 - 56404526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324.2 NP_001315.1 1 - 2 198 84
GenBank mRNA AA037145.1 - 1 - 2 199 85
AK312774.1 - 1 - 2 71 84
BC001011.2 - 1 - 2 59 84
BC007425.2 - 1 - 2 59 84
BU189971.1 - 1 - 2 164 84
L02547.1 - 1 - 2 151 84

View Full Product Details