Assay Design
Gene Transcripts
- Gene Symbol
- ATP1B4
- Entrez Gene ID
-
23439
- Gene Name
- ATPase Na+/K+ transporting family member beta 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 120362085 - 120383249 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012069.4 |
NP_036201.1 |
2 - 3 |
407 |
82 |
GenBank mRNA |
AF158384.2 |
- |
2 - 3 |
380 |
82 |
View Full Product Details