Assay Design



Gene Transcripts

Gene Symbol
SLC17A3
Entrez Gene ID
10786
Gene Name
solute carrier family 17 member 3
Gene Aliases
GOUT4, NPT4, UAQTL4
Chromosome Location
Chr.6: 25845100 - 25874243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098486.1 NP_001091956.1 5 - 6 732 77
NM_006632.3 NP_006623.2 4 - 5 498 77
GenBank mRNA AK296070.1 - 3 - 4 456 77
AK296082.1 - 4 - 5 688 77
AK298271.1 - 5 - 6 732 77
AK298367.1 - 4 - 5 495 77
BC017952.1 - 4 - 5 467 77
U90545.1 - 4 - 5 707 77
Z83953.1 - 2 - 3 321 77

View Full Product Details