Assay Design



Gene Transcripts

Gene Symbol
ARMCX3
Entrez Gene ID
51566
Gene Name
armadillo repeat containing, X-linked 3
Gene Aliases
ALEX3, GASP6, dJ545K15.2
Chromosome Location
Chr.X: 101623127 - 101631910 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016607.3 NP_057691.1 2 - 3 253 77
NM_177947.2 NP_808816.1 2 - 3 225 77
NM_177948.2 NP_808817.1 2 - 3 176 77
XM_005262141.2 XP_005262198.1 2 - 3 382 77
GenBank mRNA AB039669.1 - 2 - 3 200 77
AK222655.1 - 2 - 3 210 77
AK291225.1 - 2 - 3 254 77
AL708980.1 - 2 - 3 224 77
BC005194.1 - 2 - 3 169 77
BM905704.1 - 2 - 3 211 77
BU838618.1 - - 175 77
BX647802.1 - 2 - 3 194 77
CA397688.1 - 2 - 3 253 77
CR933656.1 - 2 - 3 347 77

View Full Product Details