Assay Design



Gene Transcripts

Gene Symbol
ENPEP
Entrez Gene ID
2028
Gene Name
glutamyl aminopeptidase
Gene Aliases
APA, CD249, gp160
Chromosome Location
Chr.4: 110476073 - 110563337 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001977.3 NP_001968.3 9 - 10 1917 151
XM_017007877.1 XP_016863366.1 6 - 7 595 151
GenBank mRNA AK313412.1 - 10 - 11 1579 151
AM888273.1 - 9 - 10 1917 151
AM888274.1 - 9 - 10 1917 151
AM888275.1 - 9 - 10 1917 151
AM888276.1 - 9 - 10 1917 151
AM888277.1 - 9 - 10 1917 151
AM888278.1 - 9 - 10 1917 151
AM888279.1 - 9 - 10 1917 151
AM888280.1 - 9 - 10 1917 151
AM888281.1 - 9 - 10 1917 151
AM888282.1 - 9 - 10 1917 151
AM888283.1 - 9 - 10 1917 151
BC094770.1 - 10 - 11 1898 151
BX641012.1 - 6 - 7 1084 151
L12468.1 - 10 - 11 1821 151
L14721.1 - 10 - 11 1658 151

View Full Product Details