Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166010.1 | NP_001159482.1 | 3 - 4 | 353 | 87 |
NM_006117.2 | NP_006108.2 | 3 - 4 | 341 | 87 | |
NM_206836.2 | NP_996667.2 | 3 - 4 | 351 | 87 | |
NR_028588.1 | 3 - 4 | 341 | 87 | ||
XM_006714957.1 | XP_006715020.1 | 3 - 4 | 344 | 87 | |
XM_017010170.1 | XP_016865659.1 | 3 - 4 | 335 | 87 | |
GenBank mRNA | AB209917.1 | - | 2 - 3 | 335 | 87 |
AF069301.1 | - | 3 - 4 | 301 | 87 | |
AF153612.1 | - | 3 - 4 | 311 | 87 | |
AF244138.1 | - | 3 - 4 | 306 | 87 | |
AF257175.1 | - | 3 - 4 | 303 | 87 | |
AK075108.1 | - | 3 - 4 | 319 | 87 | |
AK222617.1 | - | 3 - 4 | 327 | 87 | |
AK223003.1 | - | 3 - 4 | 329 | 87 | |
AK297049.1 | - | 3 - 4 | 319 | 87 | |
AY297537.1 | - | 2 - 3 | 256 | 87 | |
BC002668.2 | - | 3 - 4 | 328 | 87 | |
BC016781.1 | - | 3 - 4 | 320 | 87 | |
BC017474.1 | - | 3 - 4 | 305 | 87 | |
BC025287.1 | - | 3 - 4 | 304 | 87 | |
BC033841.1 | - | 3 - 4 | 311 | 87 | |
BC034702.1 | - | 3 - 4 | 353 | 87 | |
CR456768.1 | - | 2 - 3 | 224 | 87 |