Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128615.1 | NP_001122087.1 | 10 - 11 | 1070 | 82 |
NM_001128616.1 | NP_001122088.1 | 7 - 8 | 1486 | 82 | |
NM_001289698.1 | NP_001276627.1 | 8 - 9 | 994 | 82 | |
NM_019555.2 | NP_062455.1 | 7 - 8 | 1033 | 82 | |
XM_005265186.4 | XP_005265243.1 | 10 - 11 | 1134 | 82 | |
XM_005265187.3 | XP_005265244.1 | 10 - 11 | 1414 | 82 | |
XM_011533764.1 | XP_011532066.1 | 9 - 10 | 993 | 82 | |
XM_011533765.1 | XP_011532067.1 | 9 - 10 | 981 | 82 | |
XM_011533766.2 | XP_011532068.1 | 9 - 10 | 1325 | 82 | |
XM_011533767.1 | XP_011532069.1 | 8 - 9 | 945 | 82 | |
XM_017006502.1 | XP_016861991.1 | 12 - 13 | 1335 | 82 | |
XM_017006503.1 | XP_016861992.1 | 10 - 11 | 1070 | 82 | |
XM_017006504.1 | XP_016861993.1 | 10 - 11 | 1128 | 82 | |
XM_017006505.1 | XP_016861994.1 | 7 - 8 | 1025 | 82 | |
XM_017006506.1 | XP_016861995.1 | 7 - 8 | 1287 | 82 | |
GenBank mRNA | AB209661.1 | - | 7 - 8 | 1426 | 82 |
AF249744.1 | - | 7 - 8 | 991 | 82 | |
AF433662.1 | - | 10 - 11 | 1027 | 82 | |
AK024340.1 | - | 5 - 6 | 655 | 82 | |
AK225688.1 | - | 7 - 8 | 1015 | 82 | |
AK291412.1 | - | 7 - 8 | 1015 | 82 | |
AK293864.1 | - | 7 - 8 | 1231 | 82 | |
AK300045.1 | - | 8 - 9 | 994 | 82 | |
AK302493.1 | - | 10 - 11 | 1070 | 82 | |
AL136832.1 | - | 7 - 8 | 968 | 82 | |
AX775923.1 | - | 7 - 8 | 968 | 82 | |
BC054345.1 | - | 7 - 8 | 981 | 82 | |
BC068513.1 | - | 7 - 8 | 1015 | 82 | |
BC098122.1 | - | 7 - 8 | 986 | 82 | |
BC098272.1 | - | 7 - 8 | 986 | 82 | |
BC099715.1 | - | 6 - 7 | 909 | 82 | |
BC104723.1 | - | 7 - 8 | 986 | 82 |