Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032251.5 | NP_115627.6 | 25 - 26 | 4353 | 86 |
XM_006718519.3 | XP_006718582.1 | 25 - 26 | 4223 | 86 | |
XM_011544957.2 | XP_011543259.1 | 24 - 25 | 4461 | 86 | |
XM_011544959.2 | XP_011543261.1 | 24 - 25 | 4603 | 86 | |
XM_011544960.2 | XP_011543262.1 | 24 - 25 | 4127 | 86 | |
XM_011544962.2 | XP_011543264.1 | 11 - 12 | 2083 | 86 | |
GenBank mRNA | AK074126.1 | - | 18 - 19 | 4007 | 86 |
AK090436.1 | - | 23 - 24 | 4677 | 86 | |
AL136799.1 | - | 2 - 3 | 314 | 86 | |
AY491401.1 | - | 8 - 9 | 1074 | 86 | |
BC030194.1 | - | 3 - 4 | 380 | 86 | |
BC040232.1 | - | 12 - 13 | 1768 | 86 | |
BC140822.1 | - | 25 - 26 | 4342 | 86 | |
BC140824.1 | - | 25 - 26 | 4342 | 86 |