Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032251.5 | NP_115627.6 | 16 - 17 | 2910 | 78 |
XM_006718519.3 | XP_006718582.1 | 16 - 17 | 2922 | 78 | |
XM_011544957.2 | XP_011543259.1 | 16 - 17 | 2923 | 78 | |
XM_011544959.2 | XP_011543261.1 | 16 - 17 | 2923 | 78 | |
XM_011544962.2 | XP_011543264.1 | 3 - 4 | 545 | 78 | |
GenBank mRNA | AK074126.1 | - | 10 - 11 | 2344 | 78 |
AK090436.1 | - | 16 - 17 | 2905 | 78 | |
BC040232.1 | - | 3 - 4 | 467 | 78 | |
BC140822.1 | - | 16 - 17 | 2899 | 78 | |
BC140824.1 | - | 16 - 17 | 2899 | 78 | |
BC141866.1 | - | 16 - 17 | 2889 | 78 |