Assay Design
Gene Transcripts
- Gene Symbol
- COL17A1
- Entrez Gene ID
-
1308
- Gene Name
- collagen type XVII alpha 1
- Gene Aliases
-
BA16H23.2, BP180, BPA-2, BPAG2, ERED, LAD-1
- Chromosome Location
-
Chr.10: 104031288 - 104085880 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_000494.3 |
NP_000485.3 |
36 - 37 |
2714 |
104 |
GenBank mRNA |
AB900157.1 |
- |
35 - 36 |
2545 |
104 |
M91669.1 |
- |
36 - 37 |
2650 |
104 |
View Full Product Details