Assay Design



Gene Transcripts

Gene Symbol
COL17A1
Entrez Gene ID
1308
Gene Name
collagen type XVII alpha 1
Gene Aliases
BA16H23.2, BP180, BPA-2, BPAG2, ERED, LAD-1
Chromosome Location
Chr.10: 104031288 - 104085880 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000494.3 NP_000485.3 43 - 44 98
GenBank mRNA AB900157.1 - 42 - 43 2893 98
M91669.1 - 43 - 44 2998 98

View Full Product Details