Assay Design



Gene Transcripts

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location
Chr.10: 10462515 - 11336675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025076.2 NP_001020247.1 8 - 9 1055 57
NM_001025077.2 NP_001020248.1 8 - 9 912 57
NM_001083591.1 NP_001077060.1 8 - 9 1055 57
NM_001326317.1 NP_001313246.1 10 - 11 914 57
NM_001326318.1 NP_001313247.1 10 - 11 914 57
NM_001326319.1 NP_001313248.1 11 - 12 952 57
NM_001326320.1 NP_001313249.1 11 - 12 1008 57
NM_001326321.1 NP_001313250.1 10 - 11 914 57
NM_001326323.1 NP_001313252.1 11 - 12 1008 57
NM_001326324.1 NP_001313253.1 9 - 10 891 57
NM_001326325.1 NP_001313254.1 10 - 11 985 57
NM_001326326.1 NP_001313255.1 9 - 10 910 57
NM_001326327.1 NP_001313256.1 9 - 10 910 57
NM_001326328.1 NP_001313257.1 10 - 11 1004 57
NM_001326329.1 NP_001313258.1 8 - 9 944 57
NM_001326330.1 NP_001313259.1 8 - 9 944 57
NM_001326331.1 NP_001313260.1 8 - 9 1096 57
NM_001326332.1 NP_001313261.1 8 - 9 1096 57
NM_001326333.1 NP_001313262.1 7 - 8 1016 57
NM_001326334.1 NP_001313263.1 9 - 10 1281 57
NM_001326335.1 NP_001313264.1 8 - 9 1096 57
NM_001326336.1 NP_001313265.1 8 - 9 1096 57
NM_001326337.1 NP_001313266.1 8 - 9 1096 57
NM_001326338.1 NP_001313267.1 7 - 8 1002 57
NM_001326339.1 NP_001313268.1 7 - 8 1002 57
NM_001326340.1 NP_001313269.1 8 - 9 1008 57
NM_001326341.1 NP_001313270.1 8 - 9 1008 57
NM_001326342.1 NP_001313271.1 8 - 9 1008 57
NM_001326343.1 NP_001313272.1 8 - 9 1008 57
NM_001326344.1 NP_001313273.1 8 - 9 798 57
NM_001326345.1 NP_001313274.1 8 - 9 876 57
NM_001326346.1 NP_001313275.1 6 - 7 1041 57
NM_001326347.1 NP_001313276.1 8 - 9 1206 57
NM_001326348.1 NP_001313277.1 8 - 9 1204 57
NM_001326349.1 NP_001313278.1 8 - 9 1204 57
NM_006561.3 NP_006552.3 8 - 9 1001 57
XM_005252354.4 XP_005252411.1 8 - 9 1091 57
XM_005252357.4 XP_005252414.1 6 - 7 710 57
XM_005252358.3 XP_005252415.1 6 - 7 1051 57
XM_006717373.2 XP_006717436.1 7 - 8 1131 57
XM_011519294.2 XP_011517596.1 8 - 9 1091 57
XM_011519297.1 XP_011517599.1 8 - 9 1214 57
XM_017015542.1 XP_016871031.1 7 - 8 767 57
XM_017015543.1 XP_016871032.1 8 - 9 857 57
XM_017015544.1 XP_016871033.1 8 - 9 857 57
XM_017015545.1 XP_016871034.1 8 - 9 857 57
XM_017015546.1 XP_016871035.1 8 - 9 857 57
XM_017015547.1 XP_016871036.1 8 - 9 1091 57
XM_017015548.1 XP_016871037.1 8 - 9 857 57
XM_017015549.1 XP_016871038.1 8 - 9 857 57
XM_017015550.1 XP_016871039.1 8 - 9 1837 57
XM_017015551.1 XP_016871040.1 8 - 9 1091 57
XM_017015552.1 XP_016871041.1 8 - 9 1214 57
XM_017015553.1 XP_016871042.1 8 - 9 1214 57
XM_017015554.1 XP_016871043.1 8 - 9 1214 57
XM_017015555.1 XP_016871044.1 7 - 8 767 57
XM_017015556.1 XP_016871045.1 7 - 8 1131 57
XM_017015557.1 XP_016871046.1 9 - 10 796 57
XM_017015558.1 XP_016871047.1 7 - 8 783 57
XM_017015559.1 XP_016871048.1 7 - 8 1131 57
XM_017015560.1 XP_016871049.1 7 - 8 792 57
XM_017015561.1 XP_016871050.1 7 - 8 1119 57
XM_017015562.1 XP_016871051.1 7 - 8 1131 57
XM_017015563.1 XP_016871052.1 7 - 8 1118 57
XM_017015564.1 XP_016871053.1 6 - 7 707 57
XM_017015565.1 XP_016871054.1 6 - 7 710 57
XM_017015566.1 XP_016871055.1 6 - 7 707 57
XM_017015567.1 XP_016871056.1 7 - 8 847 57
XM_017015568.1 XP_016871057.1 9 - 10 834 57
XM_017015569.1 XP_016871058.1 7 - 8 1134 57
XM_017015570.1 XP_016871059.1 6 - 7 710 57
XM_017015571.1 XP_016871060.1 7 - 8 1134 57
XM_017015572.1 XP_016871061.1 7 - 8 835 57
GenBank mRNA AF036956.1 - 8 - 9 971 57
AF090693.1 - 8 - 9 887 57
AF090694.1 - 8 - 9 839 57
AF432906.1 - 8 - 9 907 57
AK293164.1 - 8 - 9 948 57
AK295511.1 - 6 - 7 642 57
AK297313.1 - 6 - 7 832 57
AK299548.1 - 8 - 9 831 57
AK299983.1 - 7 - 8 1040 57
AK299992.1 - 8 - 9 988 57
AK302681.1 - 5 - 6 606 57
AK314087.1 - 8 - 9 1010 57
AK316354.1 - 8 - 9 807 57
BC036391.1 - 8 - 9 973 57
BM699598.1 - 2 - 3 96 57
BP362205.1 - 6 - 7 553 57
U69546.1 - 8 - 9 788 57

View Full Product Details