Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025065.6 | NP_079341.2 | 6 - 7 | 745 | 127 |
GenBank mRNA | AF322053.1 | - | 6 - 7 | 700 | 127 |
AK022537.1 | - | 6 - 7 | 716 | 127 | |
AL359584.2 | - | 4 - 5 | 422 | 127 | |
BC016051.1 | - | 6 - 7 | 707 | 127 | |
CR627468.1 | - | 4 - 5 | 422 | 127 |