Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025065.6 | NP_079341.2 | 7 - 8 | 926 | 103 |
GenBank mRNA | AA761809.1 | - | 2 - 3 | 103 | |
AF322053.1 | - | 7 - 8 | 881 | 103 | |
AK022537.1 | - | 7 - 8 | 897 | 103 | |
AL359584.2 | - | 5 - 6 | 603 | 103 | |
BC016051.1 | - | 7 - 8 | 888 | 103 | |
CR627468.1 | - | 5 - 6 | 603 | 103 |