Assay Design
Gene Transcripts
- Gene Symbol
- CLCA3P
- Entrez Gene ID
-
9629
- Gene Name
- chloride channel accessory 3, pseudogene
- Gene Aliases
-
CLCA3
- Chromosome Location
-
Chr.1: 86634276 - 86655376 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024604.1 |
|
12 - 13 |
|
134 |
GenBank mRNA |
AF043976.1 |
- |
12 - 13 |
2036 |
134 |
View Full Product Details